Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
Thermo Scientific Chemicals
Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent
CAS: 10378-47-9 | H16CeN4O16S4·2H2O | 632.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass Bottle
Catalog number FSA325795000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium cerium(IV) sulfate dihydrate
Melting Point130.0°C
CAS10378-47-9
ColorOrange
Health Hazard 1GHS Signal Word: Warning