Methanol, 99.9%, for HPLC gradient grade
Methanol, 99.9%, for HPLC gradient grade
Methanol, 99.9%, for HPLC gradient grade
Thermo Scientific Chemicals
Methanol, 99.9%, for HPLC gradient grade
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA325740025
Price (MYR)
517.00
Bottle
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
517.00
Bottle
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 250nm, 0.005 max. at 260nm, 0.01 max. at 240nm, 0.05 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm
CAS67-56-1
ColorColorless
View more
For HPLC
RUO – Research Use Only