(-)-Caryophyllene oxide, 95%
(-)-Caryophyllene oxide, 95%
(-)-Caryophyllene oxide, 95%
(-)-Caryophyllene oxide, 95%
Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%
CAS: 1139-30-6 | C15H24O | 220.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA325080500
Price (MYR)
632.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
632.00
BOT
Add to cart
Specifications
Chemical Name or Material(-)-Caryophyllene oxide
CAS1139-30-6
CAS Min %94.0
CAS Max %100.0
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.