Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals
Deuterium oxide, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
CAS: 7789-20-0 | H2O | 20.03 g/mol
Have Questions?
Catalog NumberQuantity
FSA3207100757.5 mL
Catalog number FSA320710075
Price (MYR)
262.00
BOT
Add to cart
Quantity:
7.5 mL
Request bulk or custom format
Price (MYR)
262.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDeuterium oxide
Density1.1050g/mL
Deuterium (D)99.85% min.
Physical FormLiquid
Refractive Index1.328
View more
For NMR
RUO – Research Use Only