Tris(dibenzylideneacetone)dipalladium(0), 97%
Tris(dibenzylideneacetone)dipalladium(0), 97%
Tris(dibenzylideneacetone)dipalladium(0), 97%
Tris(dibenzylideneacetone)dipalladium(0), 97%
Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%
CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number FSA318775000
Price (MYR)
485.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
485.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTris(dibenzylideneacetone)dipalladium(0)
CAS51364-51-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.