Cobalt(II) bromide hydrate
Cobalt(II) bromide hydrate
Cobalt(II) bromide hydrate
Cobalt(II) bromide hydrate
Thermo Scientific Chemicals
Cobalt(II) bromide hydrate
CAS: 85017-77-2 | Br2Co | 218.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA318541000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II) bromide hydrate
CAS85017-77-2
ColorViolet
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.