Copper, 99+%, turnings
Copper, 99+%, turnings
Copper, 99+%, turnings
Copper, 99+%, turnings
Thermo Scientific Chemicals
Copper, 99+%, turnings
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA317912500
Price (MYR)
352.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
352.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCopper
Name Note99+%
CAS7440-50-8
Melting Point1083.0°C
Boiling Point2595.0°C