Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
CAS: 13759-83-6 | H12N3O15Sm | 444.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA316151000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSamarium(III) nitrate hexahydrate
CAS13759-83-6
ColorYellow
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May intensify fire; oxidizer.
View more
For ultra trace analysis
RUO – Research Use Only