Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)
CAS: 10031-51-3 | ErH10N3O14 | 443.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA315141000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialErbium(III) nitrate pentahydrate
CAS10031-51-3
ColorPink
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
For ultra trace analysis
RUO – Research Use Only