3-(chloromethyl)benzoyl chloride, 98%
3-(chloromethyl)benzoyl chloride, 98%
3-(chloromethyl)benzoyl chloride, 98%
3-(chloromethyl)benzoyl chloride, 98%
Thermo Scientific Chemicals
3-(chloromethyl)benzoyl chloride, 98%
CAS: 63024-77-1 | C8H6Cl2O | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA314430050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-(chloromethyl)benzoyl chloride
CAS63024-77-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0