Pentyl isocyanate, 98%
Pentyl isocyanate, 98%
Pentyl isocyanate, 98%
Pentyl isocyanate, 98%
Thermo Scientific Chemicals
Pentyl isocyanate, 98%
CAS: 3954-13-0 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA314320050
Price (MYR)
726.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
726.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPentyl isocyanate
Name Note98%
CAS3954-13-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if inhaled. May cause respiratory irritation. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Flammable liquid and vapor.