3,5-Dichlorophenyl isocyanate, 96%
3,5-Dichlorophenyl isocyanate, 96%
3,5-Dichlorophenyl isocyanate, 96%
3,5-Dichlorophenyl isocyanate, 96%
Thermo Scientific Chemicals
3,5-Dichlorophenyl isocyanate, 96%
CAS: 34893-92-0 | C7H3Cl2NO | 188.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA314140250
Price (MYR)
1,206.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,206.00
EA
Add to cart
Specifications
Chemical Name or Material3, 5-Dichlorophenyl isocyanate
CAS34893-92-0
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0