Benzyl phenyl ether, 98%
Benzyl phenyl ether, 98%
Benzyl phenyl ether, 98%
Benzyl phenyl ether, 98%
Benzyl phenyl ether, 98%
CAS: 946-80-5 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA310260250
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzyl phenyl ether
CAS946-80-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0