D-Glucuronic acid, 98%
D-Glucuronic acid, 98%
D-Glucuronic acid, 98%
D-Glucuronic acid, 98%
Thermo Scientific Chemicals
D-Glucuronic acid, 98%
CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Packaging:
Plastic bottle
Catalog number FSA309880250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Glucuronic acid
Name Note98%
CAS6556-12-3
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.