D-Trehalose, 99%, anhydrous
D-Trehalose, 99%, anhydrous
D-Trehalose, 99%, anhydrous
D-Trehalose, 99%, anhydrous
Thermo Scientific Chemicals
D-Trehalose, 99%, anhydrous
CAS: 99-20-7 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA309870250
Price (MYR)
742.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
742.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD-Trehalose
Name NoteAnhydrous
CAS99-20-7
Health Hazard 2GHS H Statement:
Causes skin irritation.
Health Hazard 3GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.