Tin(II) fluoride, 99%
Tin(II) fluoride, 99%
Tin(II) fluoride, 99%
Tin(II) fluoride, 99%
Thermo Scientific Chemicals
Tin(II) fluoride, 99%
CAS: 7783-47-3 | F2Sn | 156.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA308501000
Price (MYR)
555.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
555.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTin(II) fluoride
CAS7783-47-3
ColorWhite
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement