4-Vinylaniline, 90%, technical, stabilized
4-Vinylaniline, 90%, technical, stabilized
4-Vinylaniline, 90%, technical, stabilized
4-Vinylaniline, 90%, technical, stabilized
Thermo Scientific Chemicals
4-Vinylaniline, 90%, technical, stabilized
CAS: 1520-21-4 | C8H9N | 119.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA308480050
Price (MYR)
1,133.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,133.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Vinylaniline
CAS1520-21-4
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0