L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals
L-Alanine methyl ester hydrochloride, 99%
CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA308360050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Alanine methyl ester hydrochloride
Name Note99%
CAS2491-20-5
Health Hazard 1HYGROSCOPIC
Melting Point109.0°C to 111.0°C