3,5-Bis(trifluoromethyl)benzaldehyde, 97%
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
CAS: 401-95-6 | C9H4F6O | 242.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA307690050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 5-Bis(trifluoromethyl)benzaldehyde
CAS401-95-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0