6-Methylnicotinic acid, 99%
6-Methylnicotinic acid, 99%
6-Methylnicotinic acid, 99%
6-Methylnicotinic acid, 99%
Thermo Scientific Chemicals
6-Methylnicotinic acid, 99%
CAS: 3222-47-7 | C7H7NO2 | 137.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA307620050
Price (MYR)
774.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
774.00
EA
Add to cart
Specifications
Chemical Name or Material6-Methylnicotinic acid
CAS3222-47-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0