Sodium thiomethoxide, 95%, pure
Sodium thiomethoxide, 95%, pure
Sodium thiomethoxide, 95%, pure
Sodium thiomethoxide, 95%, pure
Thermo Scientific Chemicals
Sodium thiomethoxide, 95%, pure
CAS: 5188-07-8 | CH3NaS | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA305710050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium thiomethoxide
CAS5188-07-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta