Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg | CAS: 18154-32-0 | C10H12FeN2NaO8·3H2O | 421.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number FSA304682500
Price (MYR)
611.00
BOT
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
611.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid, ferric sodium salt trihydrate
Name Note98%
CAS18154-32-0
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Chelation/Complexation reagent
RUO – Research Use Only