4-Bromophenol, 97%
4-Bromophenol, 97%
4-Bromophenol, 97%
Thermo Scientific Chemicals
4-Bromophenol, 97%
CAS: 106-41-2 | C6H5BrO | 173.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA304411000
Price (MYR)
448.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
448.00
EA
Add to cart
Specifications
Chemical Name or Material4-Bromophenol
CAS106-41-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0