Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Thermo Scientific Chemicals
Palmitoyl chloride, 98%
CAS: 112-67-4 | C16H31ClO | 274.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA304291000
Price (MYR)
416.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
416.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPalmitoyl chloride
Name Note98%
Acidity1% max. (as palmitic acid)
CAS112-67-4
Health Hazard 1GHS Signal Word: Danger