Metacycline hydrochloride, 98%, sec. Standard
Metacycline hydrochloride, 98%, sec. Standard
Metacycline hydrochloride, 98%, sec. Standard
Metacycline hydrochloride, 98%, sec. Standard
Metacycline hydrochloride, 98%, sec. Standard
Have Questions?
Catalog NumberQuantityPackaging
FSA303861000100 mgGlass Bottle
Catalog number FSA303861000
Price (MYR)
1,032.00
MG
Add to cart
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,032.00
MG
Add to cart
Specifications
Chemical Name or MaterialMetacycline hydrochloride
Name Note98%
CAS3963-95-9
Health Hazard 1LIGHT SENSITIVE
ColorYellow