4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzoyl chloride, 97%
CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA303420250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzoyl chloride
CAS329-15-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0