D-Arabitol, 99%
D-Arabitol, 99%
D-Arabitol, 99%
Thermo Scientific Chemicals
D-Arabitol, 99%
CAS: 488-82-4 | C5H12O5 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA302870250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Arabitol
CAS488-82-4
Melting Point97°C to 105°C
ColorWhite
EINECS Number207-686-2