4,6-Dimethyldibenzothiophene, 95%
4,6-Dimethyldibenzothiophene, 95%
4,6-Dimethyldibenzothiophene, 95%
4,6-Dimethyldibenzothiophene, 95%
Thermo Scientific Chemicals
4,6-Dimethyldibenzothiophene, 95%
CAS: 1207-12-1 | C14H12S | 212.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA302190050
Price (MYR)
3,290.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,290.00
EA
Add to cart
Specifications
Chemical Name or Material4, 6-Dimethyldibenzothiophene
CAS1207-12-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0