Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
Thermo Scientific Chemicals
Sulfuric acid, pure, 96.00% (+/- 0.50%) solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3020700252.5 LGlass bottle
Catalog number FSA302070025
Price (MYR)
464.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
464.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfuric acid
ColorColorless
Name Note96.00% (±0.50%) Solution in Water
CAS7664-93-9
Melting Point10.0°C