Diphenyl ditelluride, 97%
Diphenyl ditelluride, 97%
Diphenyl ditelluride, 97%
Diphenyl ditelluride, 97%
Thermo Scientific Chemicals
Diphenyl ditelluride, 97%
CAS: 32294-60-3 | C12H10Te2 | 409.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA301840010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiphenyl ditelluride
CAS32294-60-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge