2-Hydroxybenzylamine, 98%
2-Hydroxybenzylamine, 98%
2-Hydroxybenzylamine, 98%
2-Hydroxybenzylamine, 98%
Thermo Scientific Chemicals
2-Hydroxybenzylamine, 98%
CAS: 932-30-9 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA298890010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Hydroxybenzylamine
Name Note98%
CAS932-30-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning