1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
Thermo Scientific Chemicals
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
CAS: 41100-52-1 | C12H22ClN | 215.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA298080010
Price (MYR)
432.00
G
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
432.00
G
Add to cart
Specifications
Chemical Name or Material1, 3-Dimethyl-5-aminoadamantane hydrochloride
CAS41100-52-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec