4-(Trifluoromethoxy)aniline, 99%
4-(Trifluoromethoxy)aniline, 99%
4-(Trifluoromethoxy)aniline, 99%
4-(Trifluoromethoxy)aniline, 99%
Thermo Scientific Chemicals
4-(Trifluoromethoxy)aniline, 99%
CAS: 461-82-5 | C7H6F3NO | 177.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
2.5 g
Packaging:
Glass bottle
Catalog number FSA297640250
Price (MYR)
1,087.00
G
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,087.00
G
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethoxy)aniline
Name Note99+%
CAS461-82-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger