Camphene, 75%, remainder mainly alpha-fenchene
Camphene, 75%, remainder mainly alpha-fenchene
Camphene, 75%, remainder mainly alpha-fenchene
Camphene, 75%, remainder mainly alpha-fenchene
Thermo Scientific Chemicals
Camphene, 75%, remainder mainly alpha-fenchene
CAS: 79-92-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA295570010
Price (MYR)
680.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
680.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCamphene
Name Note75%
CAS79-92-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Flammable solid.
Very toxic to aquatic life with long lasting effects.