Check Order Status
7-Methoxy-2-tetralone, 95%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
7-Methoxy-2-tetralone, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number
FSA294970050
Price (MYR)
460.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
460.00
EA
Add to cart
Specifications
Chemical Name or Material
7-Methoxy-2-tetralone
CAS
4133-34-0
CAS Min %
94.0
CAS Max %
100.0
Color
Yellow to Orange