4-Quinolinecarboxaldehyde, 95%
4-Quinolinecarboxaldehyde, 95%
4-Quinolinecarboxaldehyde, 95%
4-Quinolinecarboxaldehyde, 95%
Thermo Scientific Chemicals
4-Quinolinecarboxaldehyde, 95%
CAS: 4363-93-3 | C10H7NO | 157.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA294820050
Price (MYR)
879.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
879.00
EA
Add to cart
Specifications
Chemical Name or Material4-Quinolinecarboxaldehyde
Name Note95%
CAS4363-93-3
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning