4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
Thermo Scientific Chemicals
4-Methoxybenzylchloride, 98%, stabilized
CAS: 824-94-2 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA294171000
Price (MYR)
2,357.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,357.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methoxybenzylchloride
Name Note98%
CAS824-94-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger