3,5-Difluorobenzaldehyde, 98+%
3,5-Difluorobenzaldehyde, 98+%
3,5-Difluorobenzaldehyde, 98+%
3,5-Difluorobenzaldehyde, 98+%
Thermo Scientific Chemicals
3,5-Difluorobenzaldehyde, 98+%
CAS: 32085-88-4 | C7H4F2O | 142.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA293470050
Price (MYR)
635.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
635.00
EA
Add to cart
Specifications
Chemical Name or Material3, 5-Difluorobenzaldehyde
Name Note98+%
CAS32085-88-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning