4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
Thermo Scientific Chemicals
4-tert-Butylcalix[4]arene, 99%
CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA290220250
Price (MYR)
2,035.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,035.00
BOT
Add to cart
Specifications
Chemical Name or Material4-tert-Butylcalix[4]arene
CAS60705-62-6
CAS Min %98.5
CAS Max %100.0
Melting Point>300.0°C
View more
Chelation/Complexation reagent
RUO – Research Use Only