Polysorbate 60
Polysorbate 60
Polysorbate 60
Thermo Scientific Chemicals
Polysorbate 60
CAS: 9005-67-8 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | 522.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA278622500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPolysorbate 60
ColorYellow
Acid Value2mg KOH/g max.
CAS9005-67-8
Physical FormPaste at 25°C