2-Nitrobenzoic acid, 95%
2-Nitrobenzoic acid, 95%
2-Nitrobenzoic acid, 95%
2-Nitrobenzoic acid, 95%
Thermo Scientific Chemicals
2-Nitrobenzoic acid, 95%
CAS: 552-16-9 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA278320250
Price (MYR)
257.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
257.00
EA
Add to cart
Specifications
Chemical Name or Material2-Nitrobenzoic acid
Name Note85-90%
CAS552-16-9
CAS Min %85.0
Health Hazard 1GHS Signal Word: Warning