Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Thermo Scientific Chemicals
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA277851000
Price (MYR)
360.00
ML
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
360.00
ML
Add to cart
Chemical Identifiers
CAS1070-89-9, 109-99-9
Specifications
Chemical Name or MaterialSodium bis(trimethylsilyl)amide
ColorOrange to Brown
Name Note2M Solution in THF
CAS1070-89-9
Health Hazard 1Danger