Glycerol, pure, 83.5-88.5 wt% aqueous solution
Glycerol, pure, 83.5-88.5 wt% aqueous solution
Glycerol, pure, 83.5-88.5 wt% aqueous solution
Glycerol, pure, 83.5-88.5 wt% aqueous solution
Thermo Scientific Chemicals
Glycerol, pure, 83.5-88.5 wt% aqueous solution
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic bottle
Catalog number FSA277360050
Price (MYR)
1,443.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,443.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
Health Hazard 1
CAS7732-18-5
Melting Point18.0°C