1H,1H-Perfluoro-1-octanol, 98%
1H,1H-Perfluoro-1-octanol, 98%
1H,1H-Perfluoro-1-octanol, 98%
1H,1H-Perfluoro-1-octanol, 98%
Thermo Scientific Chemicals
1H,1H-Perfluoro-1-octanol, 98%
CAS: 307-30-2 | C8H3F15O | 400.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA277260050
Price (MYR)
397.00
G
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
397.00
G
Add to cart
Specifications
Chemical Name or Material1H, 1H-Perfluoro-1-octanol
CAS307-30-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0