o-Anisoyl chloride, 97%
o-Anisoyl chloride, 97%
o-Anisoyl chloride, 97%
o-Anisoyl chloride, 97%
Thermo Scientific Chemicals
o-Anisoyl chloride, 97%
CAS: 21615-34-9 | C8H7ClO2 | 170.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2761100505 gGlass bottle
Catalog number FSA276110050
Price (MYR)
341.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
341.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Anisoyl chloride
Name Note97%
CAS21615-34-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger