2,2'-Oxybis(ethylamine), 98%
2,2'-Oxybis(ethylamine), 98%
2,2'-Oxybis(ethylamine), 98%
2,2'-Oxybis(ethylamine), 98%
Thermo Scientific Chemicals
2,2'-Oxybis(ethylamine), 98%
CAS: 2752-17-2 | C4H12N2O | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA275840050
Price (MYR)
3,875.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,875.00
EA
Add to cart
Specifications
Chemical Name or Material2, 2'-Oxybis(ethylamine)
CAS2752-17-2
Health Hazard 1GHS Signal Word: Danger
ColorColorless
Density0.9610g/mL