MES, 99+%, <1% water
MES, 99+%, &lt;1% water
MES, 99+%, &lt;1% water
MES, 99+%, &lt;1% water
Thermo Scientific Chemicals
MES, 99+%, <1% water
CAS: 4432-31-9 | C6H13NO4S | 195.233 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA274142500
Price (MYR)
2,041.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,041.00
EA
Add to cart
Specifications
Chemical Name or MaterialMES
CAS4432-31-9
Melting Point>300.0°C
ColorWhite
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
View more
Biological buffer
RUO – Research Use Only