5-Hydroxyisophthalic acid, 99%
5-Hydroxyisophthalic acid, 99%
5-Hydroxyisophthalic acid, 99%
5-Hydroxyisophthalic acid, 99%
Thermo Scientific Chemicals
5-Hydroxyisophthalic acid, 99%
CAS: 618-83-7 | C8H6O5 | 182.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA271391000
Price (MYR)
768.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
768.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Hydroxyisophthalic acid
Name Note99%
CAS618-83-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning