Fehling B solution
Fehling B solution
Fehling B solution
Fehling B solution
Thermo Scientific Chemicals
Fehling B solution
CAS: 6381-59-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA270790025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS6381-59-5, 1310-73-2, 7732-18-5
Specifications
Chemical Name or MaterialFehling B solution
ColorColorless
CAS6381-59-5
Boiling Point100.0°C
Health Hazard 1GHS Signal Word: Danger