Iodine solution, according to Wijs, 0.1 mol ICl/l
Iodine solution, according to Wijs, 0.1 mol ICl/l
Iodine solution, according to Wijs, 0.1 mol ICl/l
Iodine solution, according to Wijs, 0.1 mol ICl/l
Thermo Scientific Chemicals
Iodine solution, according to Wijs, 0.1 mol ICl/l
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA270570025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIodine solution
ColorBrown
CAS7790-99-0
Boiling Point117.8°C
Health Hazard 1Danger